(R)-(-)-1-Aminopropan-2-ol, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
(R)-Isopropanolamine, (R)-(-)-1-Amino-2-propanol, (R)-1-Aminopropan-2-ol, (-)-1-Aminopropan-2-ol, (R)-(-)-Isopropanolamine, (-)-Isopropanolamine, (R)-1-Amino-2-propanol
M-Clarity™ quality level = MQ200
238856-1G
238856-5G
SIAL238856-1GEA
1810
CZK
SIAL238856-1G
SIAL238856-5G
(R)-(-)-1-Aminopropan-2-ol, Sigma-Aldrich®
(R)-(-)-1-Aminopropan-2-ol
Vzorec:
C₃H₉NO MW: 75.11 Bod varu: 160 Bod tání: 24…26 °C Hustota: 0.95 Storage Temperature: Refrigerator |
MDL Number:
MFCD00064428 CAS číslo: 2799-16-8 EINECS: 220-532-9 UN: 3259 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...