(R)-(-)-1-Aminopropan-2-ol 98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
(R)-Isopropanolamine, (R)-(-)-1-Amino-2-propanol, (R)-1-Aminopropan-2-ol, (-)-1-Aminopropan-2-ol, (R)-(-)-Isopropanolamine, (-)-Isopropanolamine, (R)-1-Amino-2-propanol
L14101.03
L14101.06
L14101.03EA
1410
CZK
L14101.03
L14101.06
(R)-(-)-1-Aminopropan-2-ol 98%
(R)-(-)-1-Aminopropan-2-ol
Purity: 98%
CAS No.: 2799-16-8
Molecular Formula: C3H9NO
Formula Weight: 75.11
Flash Point: 71°(159°F)
Refractive Index: 1.4500
Melting Point: 24-26°
Boiling Point: 160°
Beilstein Reference: 1718869
Optical Rotation: -19° (c=1.8 in water)
UN No.: 3259
MDL No.: MFCD00064428
Vzorec:
C₃H₉NO MW: 75.11 Bod varu: 160 °C Bod tání: 24…26 °C Hustota: 0.95 Bod vzplanutí: 71 °C (159 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00064428 CAS číslo: 2799-16-8 EINECS: 220-532-9 UN: 3259 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...