(R)-(-)-sec-Butylamine, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
(R)-(-)-2-Butylamine, (R)-(-)-2-Aminobutane
M-Clarity™ quality level = MQ200
296643-1G
296643-250MG
SIAL296643-1GEA
11510
CZK
SIAL296643-1G
SIAL296643-250MG
(R)-(-)-sec-Butylamine, Sigma-Aldrich®
(R)-(-)-sec-Butylamine
Vzorec:
C₄H₁₁N Bod varu: 63 °C (1013 hPa) Bod tání: –104 °C Hustota: 0,725 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00064416 CAS číslo: 13250-12-9 EINECS: 236-232-6 UN: 2733 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...