L(+)-Diamoniumtartrát ≥98%
Dodavatel: Thermo Fisher Scientific
Synonyms:
(R,R)-(+)-Diammonium tartrate, (R,R)-(+)-Ammonium tartrate dibasic, Ammonium tartrate dibasic, L(+)-Tartaric acid diammonium salt, (+)-Diammonium L-tartrate, (R,R)-(+)-Tartaric acid diammonium salt, L(+)-Ammonium tartrate dibasic, Ammonium L-(+)-tartrate
Purity: 98%
CAS No.: 3164-29-2
Molecular Formula: C4H12N2O6
Formula Weight: 184.15
Physical Form: Crystalline
Density: 1.6
Appearance: White
Odour: Odourless
Solubility: Soluble in water, alcohol
Beilstein Reference: 6120352
MDL No.: MFCD00013073
Vzorec:
C₄H₁₂N₂O₆ MW: 184.15 Hustota: 1.6 Storage Temperature: Ambient |
MDL Number:
MFCD00013073 CAS číslo: 3164-29-2 EINECS: 221-618-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...