Tisk…

L(+)-Diamoniumtartrát 99%

Dodavatel: Thermo Fisher Scientific

211810050 211812500 211810010
ACRO211810050EA 927 CZK
ACRO211810050 ACRO211812500 ACRO211810010
L(+)-Diamoniumtartrát 99%
L(+)-Diamoniumtartrát
Vzorec: C₄H₁₂N₂O₆
Hustota: 1,6 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00013073
CAS číslo: 3164-29-2
EINECS: 221-618-9

Objednat

Výsledky specifikačního testu

Appearance Semitransparent white Crystals
Infrared spectrum Conforms
Sulfated ash ≤0.05 %
pH 6.0 to 7.0 (5 % solution)
Specific optical rotation +31.4° to +33.4° (20°C, 589 nm) (c=1.84, H2O)
Titration with HClO4 ≥98.5 %
Chloride (Cl) ≤50 ppm
Sulfate (SO4) ≤50 ppm
Iron (Fe) ≤10 ppm
Heavy metals (as Pb) ≤10 ppm
Insoluble matter ≤0.005 % (in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR