Tisk…
L(+)-vinan sodno-draselný tetrahydrát, Sigma-Aldrich®
Dodavatel: Merck
Synonyms: Potassium sodium tartrate tetrahydrate, Potassium sodium (2R,3R)-2,3-dihydroxysuccinate tetrahydrate, Potassium sodium L(+)-tartrate tetrahydrate, Sodium potassium tartrate, (R,R)-(+)-Tartaric acid potassium sodium salt tetrahydrate, (R,R)-(+)-Potassium sodium tartrate tetrahydrate, Rochelle salt, L(+)-Tartaric acid potassium sodium salt tetrahydrate, (+)-Potassium sodium L-tartrate tetrahydrate
M-Clarity™ quality level = MQ200
217255-100G 217255-2.5KG 217255-500G
SIAL217255-100GEA 1330 CZK
SIAL217255-100G SIAL217255-2.5KG SIAL217255-500G
L(+)-vinan sodno-draselný tetrahydrát, Sigma-Aldrich®
L(+)-vinan sodno-draselný tetrahydrát
Vzorec: KOOC(CHOH)₂COONa·4H₂O
MW: 282.22
Bod tání: 70…80 °C
Hustota: ∼1.767 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150989
CAS číslo: 6381-59-5
EINECS: 206-156-8
Merck Index: 13,07755
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d585522f4e3b14a7b011dc8a858f5eeca413864&vid=542d22225b124dda886e5007091b35db67214bb7&prid=web&clid=SAPDAM