D-(-)-Tartaric acid ≥99%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
unnatural tartaric acid, (2S,3S)-2,3-Dihydroxysuccinic acid, (S,S)-(-)-Tartaric acid, (S,S)-(-)-2,3-Dihydroxysuccinic acid
A11264.14
A11264.22
A11264.36
A11264.14EA
1540
CZK
A11264.14
A11264.22
A11264.36
D-(-)-Tartaric acid ≥99%
D-(-)-Tartaric acid
Purity: 99%
CAS No.: 147-71-7
Molecular Formula: C4H6O6
Formula Weight: 150.09
Flash Point: 210°(410°F)
Odour: Odourless
pH: 1.6
Melting Point: 166-170°
Beilstein Reference: 1725145
Merck Reference: 14,9068
Optical Rotation: -13° (c=10 in water)
MDL No.: MFCD00004238
Vzorec:
C₄H₆O₆ Bod varu: 356 °C (1013 hPa) Bod tání: 166…170 °C Hustota: 1,76 g/cm³ (20 °C) Bod vzplanutí: 210 °C (410 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004238 CAS číslo: 147-71-7 EINECS: 205-695-6 Merck Index: 14,9068 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...