Tisk…

D-(+)-Malic acid, Sigma-Aldrich®

Dodavatel: Merck
Varování

Synonyms: (R)-(+)-Hydroxysuccinic acid, D(+)-Hydroxy-1,4-butanedioic acid, (R)-(+)-Hydroxy-1,4-butanedioic acid, D(+)-Malic acid, D(+)-Hydroxysuccinic acid, (R)-2-Hydroxysuccinic acid

M-Clarity™ quality level = MQ200

02300-1G 02300-25G 02300-5G
SIAL02300-1GEA 1120 CZK
SIAL02300-1G SIAL02300-25G SIAL02300-5G
D-(+)-Malic acid, Sigma-Aldrich®
D-(+)-Malic acid
Vzorec: C₄H₆O₅
Bod varu: 322 °C (1013 hPa)
Bod tání: 101…104 °C
Storage Temperature: Ambient
MDL Number: MFCD00004245
CAS číslo: 636-61-3
EINECS: 211-262-2
Merck Index: 13,05307

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR