Kyselina L(+)-asparágová, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
(S)-(+)-Aminosuccinic acid (S)-Aminobutanedioic acid (S)-Aminosuccinic acid H-Asp-OH L-Asparaginic acid L(+)-Aspartic acid, (S)-(+)-Aspartic acid, L(+)-2-Amino-1,4-butanedioic acid, (S)-(+)-2-Amino-1,4-butanedioic acid, L-Aspartic acid, H-Asp-OH, L-α-Aminosuccinic acid, L-αlpha-Aminosuccinic acid, (S)-2-Aminosuccinic acid, Asp, D
M-Clarity™ quality level = MQ200
A9256-100G
A9256-1KG
A9256-500G
A9256-5KG
SIALA9256-100GEA
1400
CZK
SIALA9256-100G
SIALA9256-1KG
SIALA9256-500G
SIALA9256-5KG
Kyselina L(+)-asparágová, Sigma-Aldrich®
Kyselina L(+)-asparágová
Vzorec:
C₄H₇NO₄ Bod varu: 311 °C (1013 hPa) Bod tání: 251 °C Hustota: 1,6613 g/cm³ (13 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002616 CAS číslo: 56-84-8 EINECS: 200-291-6 Merck Index: 13,00845 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...