Tisk…

D-(-)-Aspartic acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: D-Aminosuccinic acid, D(-)-2-Amino-1,4-butanedioic acid, (R)-(-)-2-Amino-1,4-butanedioic acid, (R)-(-)-Aspartic acid, (R)-()-Aminosuccinic acid, (R)-Aminosuccinic acid, (R)-2-Aminosuccinic acid, D-Asparaginic acid, D-Aspartic acid, H-D-Asp-OH

M-Clarity™ quality level = MQ100

219096-100G 219096-25G
SIAL219096-100GEA 11510 CZK
SIAL219096-100G SIAL219096-25G
D-(-)-Aspartic acid, Sigma-Aldrich®
D-(-)-Aspartic acid
Vzorec: C₄H₇NO₄
MW: 133,1 g/mol
Bod tání: 251 °C
MDL Number: MFCD00063081
CAS číslo: 1783-96-6
EINECS: 217-234-6
Merck Index: 13,00845

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR