D-(-)-Lithium lactate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
D(-)-Lactic acid lithium salt
M-Clarity™ quality level = MQ200
L1000-100MG
L1000-1G
SIALL1000-100MGEA
0
CZK
SIALL1000-100MG
SIALL1000-1G
D-(-)-Lithium lactate, Sigma-Aldrich®
D-(-)-Lithium lactate
Vzorec:
C₃H₅LiO₃ MW: 96,01 g/mol |
CAS číslo:
27848-81-3 EINECS: 248-693-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...