Tisk…

L(+)-Norleucin, Sigma-Aldrich®

Dodavatel: Merck
Varování

Synonyms: H-Nle-OH, H-NorLeu-OH, (S)-2-Aminohexanoic acid, (S)-(+)-Norleucine

M-Clarity™ quality level = MQ200

N6877-10G N6877-1G N6877-5G
SIALN6877-10GEA 19530 CZK
SIALN6877-10G SIALN6877-1G SIALN6877-5G
L(+)-Norleucin, Sigma-Aldrich®
L(+)-Norleucin
Vzorec: C₆H₁₃NO₂
MW: 131,17 g/mol
Bod varu: 246 °C (1013 hPa)
Bod tání: 113 °C
MDL Number: MFCD00064423
CAS číslo: 327-57-1
EINECS: 206-321-4
Merck Index: 13,06738

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR