Tisk…

D-(-)-Arginine hydrochloride, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: D-Arginine monohydrochloride, (R)-(-)-Arginine hydrochloride, H-D-Arg-OH.HCl

M-Clarity™ quality level = MQ200

A6757-1G A6757-5G
SIALA6757-1GEA 3960 CZK
SIALA6757-1G SIALA6757-5G
D-(-)-Arginine hydrochloride, Sigma-Aldrich®
D-(-)-Arginine hydrochloride
Vzorec: C₆H₁₅ClN₄O₂
Bod tání: 220 °C
MDL Number: MFCD00012620
CAS číslo: 627-75-8
EINECS: 211-010-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR