(-)-Dimethyl-D-tartrate ≥99%
Dodavatel: Thermo Scientific
Synonyms:
D(-)-Tartaric acid dimethyl ester, (2S,3S)-Dimethyl 2,3-dihydroxysuccinate, (S,S)-(-)-Tartaric acid dimethyl ester, (S,S)-(-)-Dimethyl tartrate, (-)-Dimethyl D-tartrate
Purity: 99%
CAS No.: 13171-64-7
Molecular Formula: C6H10O6
Formula Weight: 178.14
Flash Point:
Vzorec:
C₆H₁₀O₆ MW: 178,14 g/mol Bod varu: 158 °C (11 mmHg) Bod tání: 48…56 °C Hustota: 1,30 Bod vzplanutí: >170 °C (338 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008445 CAS číslo: 13171-64-7 EINECS: 236-118-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...