L(+)-Diethyl tartrate ≥98%
Dodavatel: Thermo Scientific
Varování
|
Synonyms:
(+)-Diethyl L-tartrate, (R,R)-(+)-Tartaric acid diethyl ester, (2R,3R)-(+)-Diethyl tartrate, Diethyl L-tartrate, L(+)-Tartaric acid diethyl ester, (2R,3R)-Diethyl 2,3-dihydroxysuccinate, (R,R)-(+)-Diethyl tartrate
Purity: 98%
CAS No.: 87-91-2
Molecular Formula: C8H14O6
Formula Weight: 206.19
Flash Point: 93°(199°F)
Refractive Index: 1.4460
Melting Point: 17°
Boiling Point: 280°
Beilstein Reference: 1727145
Merck Reference: 14,3855
Optical Rotation: +27° (c=1 in water)
MDL No.: MFCD00009143
Vzorec:
C₈H₁₄O₆ MW: 206,2 g/mol Bod varu: 280 °C Bod tání: 17 °C Hustota: 1,205 Bod vzplanutí: 93 °C (199 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009143 CAS číslo: 87-91-2 EINECS: 201-783-3 Merck Index: 14,3855 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...