L(+)-Diethyl tartrate 99+%
Dodavatel: Thermo Fisher Scientific
Synonyms:
(+)-Diethyl L-tartrate, (R,R)-(+)-Tartaric acid diethyl ester, (2R,3R)-(+)-Diethyl tartrate, Diethyl L-tartrate, L(+)-Tartaric acid diethyl ester, (2R,3R)-Diethyl 2,3-dihydroxysuccinate, (R,R)-(+)-Diethyl tartrate
168961000
168965000
ACRO168961000EA
1480
CZK
ACRO168961000
ACRO168965000
L(+)-Diethyl tartrate 99+%
L(+)-Diethyl tartrate
Vzorec:
C₈H₁₄O₆ MW: 206.2 Bod varu: 280 Bod tání: 17 °C Hustota: 1.2 g/cm³ Bod vzplanutí: 93 °C |
MDL Number:
MFCD00009143 CAS číslo: 87-91-2 EINECS: 201-783-3 Merck Index: 13,03886 |
Výsledky specifikačního testu
Appearance | Clear viscous liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4450 to 1.4470 (20°C, 589 nm) |
GC | ≥99.0 % |
Color scale | ≤50 APHA |
Specific optical rotation | +7.5° to +9° (20°C, 589 nm) (neat) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...