L(+)-Asparagin, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
L-Asparagine, (S)-(+)-Asparagine, H-Asn-OH
M-Clarity™ quality level = MQ400
A0884-100G
A0884-10MG
A0884-1KG
A0884-25G
A0884-500G
SIALA0884-100GEA
5340
CZK
SIALA0884-100G
SIALA0884-10MG
SIALA0884-1KG
SIALA0884-25G
SIALA0884-500G
L(+)-Asparagin, Sigma-Aldrich®
L(+)-Asparagin
Vzorec:
H₂NCOCH₂CH(NH₂)CO₂H MW: 132,12 g/mol Bod varu: 343 °C (1013 hPa) Bod tání: 234…235 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064401 CAS číslo: 70-47-3 EINECS: 200-735-9 Merck Index: 12,00872 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...