Tisk…

N-acetyl-L-cystein, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Acetylcysteine, N-Acetyl-3-mercaptoalanine, NAC, (R)-(+)-N-Acetylcysteine, (R)-2-Acetamido-3-mercaptopropanoic acid, Ac-Cys-OH, N-Acetyl-L-Cysteine, LNAC

M-Clarity™ quality level = MQ200

A7250-500G A7250-25G A7250-50G A7250-100G A7250-10G A7250-1KG A7250-5G
SIALA7250-500GEA 22720 CZK
SIALA7250-500G SIALA7250-25G SIALA7250-50G SIALA7250-100G SIALA7250-10G SIALA7250-1KG SIALA7250-5G
N-acetyl-L-cystein, Sigma-Aldrich®
N-acetyl-L-cystein
Vzorec: C₅H₉NO₃S
MW: 163.2
Bod varu: 371
Bod tání: 106…108 °C
Hustota: 1.48 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004880
CAS číslo: 616-91-1
EINECS: 210-498-3
Merck Index: 13,00090

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR