Tisk…

(S)-(+)-Epichlorhydrin, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: (S)-(+)-Glycerol epichlorhydrin, (S)-(+)-3-Chloro-1,2-epoxypropane, (S)-(+)-1-Chloro-2,3-epoxypropane, (2S)-2-(chloromethyl)oxirane

M-Clarity™ quality level = MQ200

540080-25G 540080-5G
SIAL540080-25GEA 10460 CZK
SIAL540080-25G SIAL540080-5G
(S)-(+)-Epichlorhydrin, Sigma-Aldrich®
(S)-(+)-Epichlorhydrin
Vzorec: ClCH₂CH(O)CH₂
MW: 92,52 g/mol
Bod varu: 113…115 °C (1013 hPa)
Bod tání: –48 °C
Hustota: 1,18 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00077760
CAS číslo: 67843-74-7
UN: 2023
ADR: 6.1,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR