(R)-(-)-Epichlorhydrin, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
(R)-(-)-Glycerol epichlorhydrin, (R)-(-)-3-Chloro-1,2-epoxypropane, (R)-(-)-1-Chloro-2,3-epoxypropane
M-Clarity™ quality level = MQ100
540072-25G
540072-5G
SIAL540072-25GEA
13910
CZK
SIAL540072-25G
SIAL540072-5G
(R)-(-)-Epichlorhydrin, Sigma-Aldrich®
(R)-(-)-Epichlorhydrin
Vzorec:
ClCH₂CH(O)CH₂ MW: 92,52 g/mol Bod varu: 115…116 °C (1013 hPa) Bod tání: –48 °C Hustota: 1,18 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00077759 CAS číslo: 51594-55-9 EINECS: 424-280-2 UN: 2023 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...