Platinium(IV) oxide monohydrate (≥75% Pt)
Dodavatel: Thermo Scientific
|
Nebezpečí
|
CAS No.: 12137-21-2
Molecular Formula: O2Pt.H2O
Formula Weight: 245.10 (227.09anhy)
Physical Form: Powder, S.A.
Hydrogenation catalyst. Suitable for the reduction of double and triple bonds, aromatics, carbonyls, nitriles, and nitro groups
Vzorec:
O₂Pt·1H₂O MW: 245,09 g/mol Bod tání: 450 °C Storage Temperature: Ambient |
MDL Number:
MFCD00066964 CAS číslo: 12137-21-2 EINECS: 215-223-0 UN: 1479 ADR: 5.1,II Merck Index: 14,7527 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...