Platinium(IV) oxide monohydrate 99.8% (trace metals basis)
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
211930010
211932500
211930050
ACRO211930010EA
6920
CZK
ACRO211930010
ACRO211932500
ACRO211930050
Platinium(IV) oxide monohydrate 99.8% (trace metals basis)
Platinium(IV) oxide monohydrate
Vzorec:
O₂Pt·1H₂O MW: 245.09 Storage Temperature: Ambient |
MDL Number:
MFCD00066964 CAS číslo: 12137-21-2 EINECS: 215-223-0 UN: 1479 ADR: 5.1 I |
Výsledky specifikačního testu
Appearance | Brown to black Powder |
Surface area | ≥80 m²/g |
Total trace metal impurities | ≤2000 ppm |
Platinum (Pt) | 77.6 to 82 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...