Tisk…
Iridium(IV) oxide dihydrate ≥99.99% (metals basis) Ir ≥ 73%, Premion®
Dodavatel: Thermo Scientific
Synonyms: Iridium dioxide dihydrate, Iridium(IV) oxide hydrate
43051.03 43051.06
43051.03EA 20420 CZK
43051.03 43051.06
Iridium(IV) oxide dihydrate ≥99.99% (metals basis) Ir ≥ 73%, Premion®
Iridium(IV) oxide dihydrate
Purity: 99.99% CAS No.: 30980-84-8 Molecular Formula: IrO2.2H2O Formula Weight: 260.23 (224.20anhy) Physical Form: Powder
Vzorec: IrO₂·2H₂O
MW: 260,25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011065
CAS číslo: 30980-84-8
EINECS: 234-743-9