Tisk…

Ruthenium(III) chloride hydrate (35 - 40% Ru)

Dodavatel: Thermo Fisher Scientific
Nebezpečí

195480010 195480050 195480250 195481000
ACRO195480010EA 2850 CZK
ACRO195480010 ACRO195480050 ACRO195480250 ACRO195481000
Ruthenium(III) chloride hydrate (35 - 40% Ru)
Ruthenium(III) chloride hydrate
Vzorec: Cl₃Ru·xH₂O
Bod tání: ? 300 °C
Hustota: 3,11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149844
CAS číslo: 14898-67-0
EINECS: 233-167-5
UN: 3260
ADR: 8,II

Objednat

Výsledky specifikačního testu

Appearance Dark brown to black Powder
Assay 35 to 40 % (Ru)
Additional info Trace analysis values are based on Ru
Calcium (Ca) ≤300 ppm
Copper (Cu) ≤300 ppm
Gold (Au) ≤300 ppm
Iridium (Ir) ≤500 ppm
Iron (Fe) ≤300 ppm
Magnesium (Mg) ≤300 ppm
Osmium (Os) ≤100 ppm
Palladium (Pd) ≤500 ppm
Platinum (Pt) ≤500 ppm
Potassium (K) ≤300 ppm
Rhodium (Rh) ≤500 ppm
Silicon (Si) ≤300 ppm
Silver (Ag) ≤300 ppm
Sodium (Na) ≤300 ppm

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR