Chlorid roditý hydrát ≥99.99% (metals basis), Premion®
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Rhodium trichloride hydrate
43702.03
43702.06
43702.03EA
42730
CZK
43702.03
43702.06
Chlorid roditý hydrát ≥99.99% (metals basis), Premion®
Chlorid roditý hydrát
Purity: 99.99%
CAS No.: 20765-98-4
Molecular Formula: RhCl3.xH2O
Formula Weight: 209.26(anhy)
Physical Form: Crystalline
Sensitivity: Hygroscopic
Appearance: Dark red
Melting Point: 100° dec.
Merck Reference: 14,8188
MDL No.: MFCD00149839
Vzorec:
Cl₃Rh·H₂O Storage Temperature: Ambient |
MDL Number:
MFCD00149839 CAS číslo: 20765-98-4 EINECS: 233-165-4 Merck Index: 14,8188 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...