Tisk…

Iridium(III) chloride hydrate, Sigma-Aldrich®

Dodavatel: Merck
Varování

Synonyms: Iridium trichloride hydrate, Iridium(III) chloride xhydrate

M-Clarity™ quality level = MQ100

206245-1G 206245-5G
SIAL206245-1GEA 5260 CZK
SIAL206245-1G SIAL206245-5G
Iridium(III) chloride hydrate, Sigma-Aldrich®
Iridium(III) chloride hydrate
Vzorec: Cl₃Ir·xH₂O
MW: 298,58 g/mol (anhy)
Hustota: 5,3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149750
CAS číslo: 14996-61-3
EINECS: 233-044-6
Merck Index: 14,05088

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR