Iron(II) bromide hydrate
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
31133.14
31133.22
31133.14EA
3010
CZK
31133.14
31133.22
Iron(II) bromide hydrate
Iron(II) bromide hydrate
CAS No.: 7789-46-0
Molecular Formula: Br2Fe.xH2O (x≈5-6)
Formula Weight: 215.67(anhy)
Physical Form: Lump
Density: 3.9
Sensitivity: Air Sensitive and Hygroscopic
Appearance: Green
Odour: Odourless
Melting Point: 27°
UN No.: 3260
MDL No.: MFCD00016081
Vzorec:
Br₂Fe·H₂O Bod tání: 27 °C Hustota: 3,9 Storage Temperature: Ambient |
MDL Number:
MFCD00016081 CAS číslo: 13463-12-2 EINECS: 232-168-8 UN: 3260 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...