Tisk…

Síran zinečnatý heptahydrát ≥99.999% (metals basis), Puratronic®

Dodavatel: Thermo Fisher Scientific
Nebezpečí

11138.09 11138.18
11138.09EA 2340 CZK
11138.09 11138.18
Síran zinečnatý heptahydrát ≥99.999% (metals basis), Puratronic®
Zinc sulphate hydrate

Purity: 99.999% CAS No.: 16788-42-4 Molecular Formula: O4SZn.xH2O Formula Weight: 161.43(anhy) Physical Form: Crystalline powder Appearance: White Odour: Odourless Melting Point: 100° Merck Reference: 14,10159 UN No.: 3077 MDL No.: MFCD00011302

Vzorec: ZnSO₄·xH₂O
MW: 161.45 g/mol (anhy)
Hustota: 1.97 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149894
CAS číslo: 16788-42-4
EINECS: 231-793-3
UN: 3077
ADR: 9 III
Merck Index: 14,10159

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR