Copper(II) sulphate hydrate ≥99.999% (metals basis), Puratronic®
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Cupric sulphate hydrate
10701.14
10701.22
10701.14EA
3850
CZK
10701.14
10701.22
Copper(II) sulphate hydrate ≥99.999% (metals basis), Puratronic®
Copper(II) sulphate hydrate
Purity: 99.999%
CAS No.: 23254-43-5
Molecular Formula: CuSO4.xH2O
Formula Weight: 159.60(anhy)
Physical Form: Crystalline Aggregates
Odour: Odourless
pH: 4
Solubility: Soluble in water. Insoluble in alcohol
Merck Reference: 14,2653
UN No.: 3077
MDL No.: MFCD00149682
Vzorec:
CuO₄S·H₂O Hustota: 2,284 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149682 CAS číslo: 23254-43-5 EINECS: 231-847-6 UN: 3077 ADR: 9,III REACH: 01-2119520566-40 Merck Index: 14,2653 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...