Copper(II) nitrate hemipentahydrate 98+% ACS
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
405855000
405850025
405851000
ACRO405855000EA
4640
CZK
ACRO405855000
ACRO405850025
ACRO405851000
Copper(II) nitrate hemipentahydrate 98+% ACS
Copper(II) nitrate hemipentahydrate
Vzorec:
Cu(NO₃)₂·2.5H₂O Bod tání: 114 °C Hustota: Min, 1 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD02253001 CAS číslo: 19004-19-4 EINECS: 221-838-5 UN: 3085 ADR: 5.1,II |
Výsledky specifikačního testu
Appearance | Blue Crystalline powder |
Titration Iodometric | 98.0 to 102.0 % |
Chloride (Cl) | ≤20 ppm |
Sulfate (SO4) | ≤100 ppm |
Calcium (Ca) | ≤50 ppm |
Iron (Fe) | ≤50 ppm |
Lead (Pb) | ≤10 ppm |
Nickel (Ni) | ≤100 ppm |
Potassium (K) | ≤50 ppm |
Sodium (Na) | ≤100 ppm |
Insoluble matter | ≤0.01 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...