Terbium(III) nitrate hydrate ≥99.9% (REO, rare earth oxide basis)
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Packaged under argon.
Purity: 99.9%
CAS No.: 94219-56-4
Molecular Formula: Tb(NO3)3.xH2O
Formula Weight: 344.94(anhy)
Physical Form: Wet Crystalline Aggregates, Packaged under argon
Appearance: White
Merck Reference: 14,9157
UN No.: 1477
MDL No.: MFCD00011257
Vzorec:
Tb(NO₃)₃·xH₂O Storage Temperature: Ambient |
MDL Number:
MFCD00011257 CAS číslo: 94219-56-4 EINECS: 233-138-7 UN: 1477 ADR: 5.1,III Merck Index: 14,9157 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...