Tisk…

Holmium trinitrate pentahydrate, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí


M-Clarity™ quality level = MQ100

325732-10G 325732-50G
SIAL325732-10GEA 5760 CZK
SIAL325732-10G SIAL325732-50G
Holmium trinitrate pentahydrate, Sigma-Aldrich®
Holmium trinitrate pentahydrate
Vzorec: HoN₃O₉·5H₂O
MW: 441.02
Bod tání: 83.74…84.62 °C
MDL Number: MFCD00149737
CAS číslo: 14483-18-2
EINECS: 233-438-8
UN: 1477
ADR: 5.1 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR