Ytterbium(III) chloride hexahydrate, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
337927-10G
337927-50G
SIAL337927-10GEA
1930
CZK
SIAL337927-10G
SIAL337927-50G
Ytterbium(III) chloride hexahydrate, Sigma-Aldrich®
Ytterbium(III) chloride hexahydrate
Vzorec:
Cl₃Yb·6H₂O MW: 387.49 Bod tání: 154 Hustota: 2.57 g/cm³ (20 °C) |
MDL Number:
MFCD00149885 CAS číslo: 10035-01-5 EINECS: 233-800-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...