Tisk…

Ytterbium(III) chloride hexahydrate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

204870-10G 204870-50G
SIAL204870-10GEA 10570 CZK
SIAL204870-10G SIAL204870-50G
Ytterbium(III) chloride hexahydrate, Sigma-Aldrich®
Ytterbium(III) chloride hexahydrate
Vzorec: Cl₃Yb·6H₂O
MW: 387,49 g/mol
Bod tání: 154 °C
Hustota: 2,57 g/cm³ (20 °C)
MDL Number: MFCD00149885
CAS číslo: 10035-01-5
EINECS: 233-800-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR