Samarium trichloride hexahydrate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Samarium(III) chloride hexahydrate, Samarium trichloride hexahydrate
M-Clarity™ quality level = MQ100
248800-10G
248800-50G
SIAL248800-10GEA
1560
CZK
SIAL248800-10G
SIAL248800-50G
Samarium trichloride hexahydrate, Sigma-Aldrich®
Samarium trichloride hexahydrate
Vzorec:
Cl₃Sm·6H₂O MW: 364.81 Hustota: 2.383 g/cm³ (25 °C) |
MDL Number:
MFCD00011235 CAS číslo: 13465-55-9 EINECS: 233-797-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...