Samarium trichloride, hydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Samarium (III) chloride
11232.06
11232.14
11232.22
11232.06EA
1630
CZK
11232.06
11232.14
11232.22
Samarium trichloride, hydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Samarium trichloride
Purity: 99.99%
CAS No.: 10361-82-7
Molecular Formula: SmCl3.xH2O
Formula Weight: 256.71(anhy)
Physical Form: Wet Crystalline Aggregates
Sensitivity: Hygroscopic
Merck Reference: 14,8347
MDL No.: MFCD00011235
Vzorec:
Cl₃Sm·xH₂O MW: 256.72 Bod tání: 110 °C (-5H₂O) Hustota: 2.383 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011235 CAS číslo: 10361-82-7 EINECS: 233-797-0 Merck Index: 14,8347 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...