Lutetium trichloride hexahydrate, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
542075-1G
542075-5G
SIAL542075-1GEA
5340
CZK
SIAL542075-1G
SIAL542075-5G
Lutetium trichloride hexahydrate, Sigma-Aldrich®
Lutetium trichloride hexahydrate
Vzorec:
Cl₃Lu·6H₂O MW: 389.42 |
MDL Number:
MFCD00011099 CAS číslo: 15230-79-2 EINECS: 233-240-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...