Erbium trichloride hydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Dodavatel: Thermo Scientific
|
Varování
|
11305.14
11305.22
11305.14EA
7520
CZK
11305.14
11305.22
Erbium trichloride hydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Erbium trichloride hydrate
Purity: 99.99%
CAS No.: 19423-85-9
Molecular Formula: ErCl3.xH2O
Formula Weight: 273.62(anhy)
Physical Form: Crystalline Aggregates
Sensitivity: Hygroscopic
MDL No.: MFCD00010989
Vzorec:
Cl₃Er·H₂O MW: 291,63 g/mol Bod tání: 168 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010989 CAS číslo: 19423-85-9 EINECS: 233-385-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...