Ytterbium(III) bromide hydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Dodavatel: Thermo Fisher Scientific
|
Varování
|
44057.09
44057.18
44057.09EA
3540
CZK
44057.09
44057.18
Ytterbium(III) bromide hydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Ytterbium(III) bromide hydrate
Purity: 99.99%
CAS No.: 15163-03-8
Molecular Formula: YbBr3.xH2O
Formula Weight: 412.77(anhy)
Physical Form: Lump
Vzorec:
Br₃Yb·H₂O MW: 430.77 Storage Temperature: Ambient |
MDL Number:
MFCD00049613 CAS číslo: 15163-03-8 EINECS: 237-350-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...