Tisk…

Tetramethylammonium hydroxide pentahydrate, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

87741-100G 87741-25G 87741-500G
SIAL87741-100GEA 6190 CZK
SIAL87741-100G SIAL87741-25G SIAL87741-500G
Tetramethylammonium hydroxide pentahydrate, Sigma-Aldrich®
Tetramethylammonium hydroxide pentahydrate
Vzorec: C₄H₁₃NO·5H₂O
MW: 181.23
Bod tání: 65…68 °C
Hustota: 1.829 g/cm³ (20 °C)
MDL Number: MFCD00149566
CAS číslo: 10424-65-4
EINECS: 200-882-9
UN: 3423
ADR: 8 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR