Kaolin, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
China Clay, Hydrated aluminum silicate, Clay
M-Clarity™ quality level = MQ200
K1512-1KG
K1512-500G
SIALK1512-1KGEA
14730
CZK
SIALK1512-1KG
SIALK1512-500G
Kaolin, Sigma-Aldrich®
Kaolin
Vzorec:
Al₂SiO₇·2H₂O Bod tání: 1760 °C Hustota: 0,5 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00062311 CAS číslo: 1332-58-7 EINECS: 310-194-1 REACH: exempt Merck Index: 13,05300 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...