Tisk…

Kaolin, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: China Clay, Hydrated aluminum silicate, Clay

M-Clarity™ quality level = MQ200

60609-1KG 60609-25KG
SIAL60609-1KGEA 1690 CZK
SIAL60609-1KG SIAL60609-25KG
Kaolin, Sigma-Aldrich®
Kaolin
Vzorec: Al₂SiO₇·2H₂O
Bod tání: 1760 °C
Hustota: 0,5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00062311
CAS číslo: 1332-58-7
EINECS: 310-194-1
REACH: exempt
Merck Index: 13,05300

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR