Tisk…

Thallium(III) nitrate trihydrate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Thallium trinitrate trihydrate

M-Clarity™ quality level = MQ100

163015-100G 163015-25G
SIAL163015-100GEA 11820 CZK
SIAL163015-100G SIAL163015-25G
Thallium(III) nitrate trihydrate, Sigma-Aldrich®
Thallium(III) nitrate trihydrate
Vzorec: N₃O₉Tl·3H₂O
MW: 444.44
Bod tání: 102…105 °C
Hustota: 3.31…3.37 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149874
CAS číslo: 13453-38-8
EINECS: 237-325-4
UN: 2727
ADR: 6.1 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR