Tisk…

Dibenzocyclooctyne-maleimide

Dodavatel: Thermo Scientific
Varování

Synonyms: Azadibenzocyclooctyne-maleimide, ADIBO-maleimide, DBCO-maleimide

J65377.MB J65377.MC
J65377.MBEA 4400 CZK
J65377.MB J65377.MC
Dibenzocyclooctyne-maleimide
Dibenzocyclooctyne-maleimide

For incorporation of ADIBO functionality into sulfhydryl containing compounds

Vzorec: C₂₅H₂₁N₃O₄
MW: 427,46 g/mol
Storage Temperature: Freezer
MDL Number: MFCD22380750
CAS číslo: 1395786-30-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR