Hydrogenfosforečnan didraselný trihydrát, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Dipotassium hydrogen phosphate trihydrate, Potassium hydrogen phosphate trihydrate, di-Potassium hydrogen phosphate trihydrate, Potassium phosphate dibasic trihydrate, di-Potassium hydrogen orthophosphate trihydrate
M-Clarity™ quality level = MQ200
P9666-100G
P9666-1KG
P9666-500G
SIALP9666-100GEA
3340
CZK
SIALP9666-100G
SIALP9666-1KG
SIALP9666-500G
Hydrogenfosforečnan didraselný trihydrát, Sigma-Aldrich®
Hydrogenfosforečnan didraselný trihydrát
Vzorec:
K₂HPO₄·3H₂O Bod varu: ∼340 °C (1013 hPa) Hustota: 2,504 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149926 CAS číslo: 16788-57-1 EINECS: 231-834-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...