Tisk…

Kyselina tetrachlorzlatitá hydrát, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: Gold(III) chloride hydrate, Hydrogen tetrachloroaurate(III) xhydrate, Hydrogen tetrachloroaurate (III) hydrate, Hydrogen tetrachloroaurate hydrate, Chloroauric acid, tetra-Chloroauric acid hydrate

M-Clarity™ quality level = MQ200

254169-500MG 254169-5G
SIAL254169-500MGEA 8780 CZK
SIAL254169-500MG SIAL254169-5G
Kyselina tetrachlorzlatitá hydrát, Sigma-Aldrich®
Kyselina tetrachlorzlatitá hydrát
Vzorec: HAuCl₄·H₂O
MW: 357,8 g/mol
Hustota: 1,169 g/cm³ (20 °C)
MDL Number: MFCD00149903
CAS číslo: 27988-77-8
EINECS: 240-948-4
UN: 3260
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR