Kyselina p-toluensulfonová monohydrát ≥98.5% ACS
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
4-Methylbenzenesulphonic acid hydrate, Toluene-4-sulphonic acid monohydrate
Purity: 98.5+%
CAS No.: 6192-52-5
Molecular Formula: C7H8O3S.H2O
Formula Weight: 190.22 (172.20anhy)
Flash Point: 150°(302°F)
Physical Form: Crystalline
Density: 1.24
Sensitivity: Hygroscopic
Appearance: White to pink
Odour: Mild
Melting Point: 103-106°(anhy)
Boiling Point: 140°/20mm
Beilstein Reference: 3568023
Merck Reference: 14,9533
UN No.: 2585
MDL No.: MFCD00142137
Vzorec:
C₇H₈O₃S·1H₂O Bod varu: 140 °C (20 mmHg) Bod tání: 103…106 °C (anhydr.) Hustota: 1,24 Bod vzplanutí: 150 °C (302 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00142137 CAS číslo: 6192-52-5 EINECS: 203-180-0 UN: 2585 ADR: 8,III Merck Index: 14,9533 |