Kyselina 5-sulfosalicylová dihydrát ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
2-Hydroxy-5-sulphobenzoic acid dihydrate
Purity: 98%
CAS No.: 5965-83-3
Molecular Formula: C7H6O6SH.2H2O
Formula Weight: 254.22 (218.19anhy)
Physical Form: Cryst powder
Sensitivity: Light Sensitive
Appearance: White
Odour: Pungent
Melting Point: 103-112°
Solubility: Soluble in water
Beilstein Reference: 650741
Merck Reference: 14,8964
UN No.: 2585
MDL No.: MFCD00007508
Vzorec:
HO₃SC₆H₃-2-(OH)CO₂H·2H₂O MW: 254,22 g/mol Bod tání: 103…112 °C Hustota: 1,695 g/cm³ (20 °C) Bod vzplanutí: 150 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149540 CAS číslo: 5965-83-3 EINECS: 202-555-6 UN: 2585 ADR: 8,III Merck Index: 14,8964 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...