Tisk…

1-Hexanesulphonic acid sodium salt monohydrate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Sodium 1-hexanesulphonate monohydrate

52865-10G 52865-250G 52865-50G
SIAL52865-10GEA 3300 CZK
SIAL52865-10G SIAL52865-250G SIAL52865-50G
1-Hexanesulphonic acid sodium salt monohydrate, Sigma-Aldrich®
1-Hexanesulphonic acid sodium salt monohydrate
Vzorec: C₆H₁₃NaO₃S·1H₂O
MW: 206,24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00149549
CAS číslo: 207300-91-2
EINECS: 220-601-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR