Tisk…
1-Hexanesulphonic acid sodium salt monohydrate ≥99%
Dodavatel: Thermo Scientific
Synonyms: Sodium 1-hexanesulphonate monohydrate
32190.09 32190.18
32190.09EA 3030 CZK
32190.09 32190.18
1-Hexanesulphonic acid sodium salt monohydrate ≥99%
1-Hexanesulphonic acid sodium salt monohydrate
Purity: 99% CAS No.: 207300-91-2 Molecular Formula: C6H13NaO3S.H2O Formula Weight: 206.24 (188.22anhy) Physical Form: Crystalline Appearance: White Beilstein Reference: 3727014 MDL No.: MFCD00149549
Vzorec: C₆H₁₃NaO₃S·1H₂O
MW: 206,24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00149549
CAS číslo: 207300-91-2
EINECS: 220-601-3